Orders LinkContactUsLinkComponent
Protector® Filtered Glove Box, Labconco | Avantor
Protector® Filtered Glove Box, Labconco
Supplier:   Labconco
Protector® Filtered Glove Box, Labconco
Supplier:  Labconco
Certificates
Protector® Filtered Glove Box, Labconco
Protector® Filtered Glove Box, Labconco
15 Options Available Below
Product Details & Documents
Protector® Filtered Glove Boxes provide inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials
These glove boxes feature particulate containment <20 nanograms per cubic meter as confirmed by Safebridge Consultants, Incorporated and also meet ISO Class 3 standards.
Informations sur les accessoires: Glove box interior shelves kit provides shelf space across the back wall. Three stainless steel storage shelves are attached to epoxy-coated steel upright supports. Shelves have adjustable height. Installation hardware and instructions are included.
Product Family Options
Product Information
  • Electrical
    Material
    Certification
    Availability
    Price
  • 89175-282
    HEPA Filtered, Single Width
    HEPA Filtered, Single Width
    115 V, 50/60 Hz (US)
    Fiberglass
    UL, ETL, CAN/CSA
    Log in for availability
    Log in for availability
    Each
    Each - $31,550.00
    $31,550.00
    Each
       $31,550.00
    La quantité minimale de commande est de 1
    Supplier Number: 5065010
  • 89175-284
    HEPA Filtered, Single Width
    HEPA Filtered, Single Width
    208/230 V, 50/60 Hz (UK)
    Fiberglass
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $31,550.00
    $31,550.00
    Each
       $31,550.00
    Supplier Number: 5065031
  • 89175-286
    HEPA Filtered, Single Width
    HEPA Filtered, Single Width
    208/230 V, 50/60 Hz (Schuko)
    Fiberglass
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $31,550.00
    $31,550.00
    Each
       $31,550.00
    Supplier Number: 5065035
  • 89175-288
    HEPA Filtered, Single Width
    HEPA Filtered, Single Width
    208/230 V, 50/60 Hz (China/Australia)
    Fiberglass
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $31,550.00
    $31,550.00
    Each
       $31,550.00
    Supplier Number: 5065040
  • 89175-290
    HEPA Filtered, Single Width
    HEPA Filtered, Single Width
    208/230 V, 60 Hz, (US)
    Fiberglass
    UL, ETL, CAN/CSA
    Log in for availability
    Log in for availability
    Each
    Each - $31,550.00
    $31,550.00
    Each
       $31,550.00
    La quantité minimale de commande est de 1
    Supplier Number: 5065045
  • 89175-292
    ULPA Filtered, Single Width
    ULPA Filtered, Single Width
    115 V, 50/60 Hz (US)
    Stainless steel
    UL, ETL, CAN/CSA
    Log in for availability
    Log in for availability
    Each
    Each - $39,520.00
    $39,520.00
    Each
       $39,520.00
    La quantité minimale de commande est de 1
    Supplier Number: 5065510
  • 89175-294
    ULPA Filtered, Single Width
    ULPA Filtered, Single Width
    208/230 V, 50/60 Hz (UK)
    Stainless steel
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $39,520.00
    $39,520.00
    Each
       $39,520.00
    Supplier Number: 5065531
  • 89175-296
    ULPA Filtered, Single Width
    ULPA Filtered, Single Width
    208/230 V, 50/60 Hz (Schuko)
    Stainless steel
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $39,520.00
    $39,520.00
    Each
       $39,520.00
    Supplier Number: 5065535
  • 89175-298
    ULPA Filtered, Single Width
    ULPA Filtered, Single Width
    208/230 V, 50/60 Hz (China/Australia)
    Stainless steel
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $39,520.00
    $39,520.00
    Each
       $39,520.00
    Supplier Number: 5065540
  • 89175-300
    ULPA Filtered, Single Width
    ULPA Filtered, Single Width
    208/230 V, 60 Hz, (US)
    Stainless steel
    UL, ETL, CAN/CSA
    Log in for availability
    Log in for availability
    Each
    Each - $39,520.00
    $39,520.00
    Each
       $39,520.00
    La quantité minimale de commande est de 1
    Supplier Number: 5065545
  • 89175-302
    ULPA Filtered, Double Width
    ULPA Filtered, Double Width
    115 V, 50/60 Hz (US)
    Stainless steel
    UL, ETL, CAN/CSA
    Log in for availability
    Log in for availability
    Each
    Each - $80,410.00
    $80,410.00
    Each
       $80,410.00
    La quantité minimale de commande est de 1
    Supplier Number: 5065512
  • 89175-304
    ULPA Filtered, Double Width
    ULPA Filtered, Double Width
    208/230 V, 50/60 Hz (UK)
    Stainless steel
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $80,410.00
    $80,410.00
    Each
       $80,410.00
    Supplier Number: 5065533
  • 89175-306
    ULPA Filtered, Double Width
    ULPA Filtered, Double Width
    208/230 V, 50/60 Hz (Schuko)
    Stainless steel
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $80,410.00
    $80,410.00
    Each
       $80,410.00
    Supplier Number: 5065537
  • 89175-308
    ULPA Filtered, Double Width
    ULPA Filtered, Double Width
    208/230 V, 50/60 Hz (China/Australia)
    Stainless steel
    CE
    Log in for availability
    Log in for availability
    Each
    Each - $80,410.00
    $80,410.00
    Each
       $80,410.00
    Supplier Number: 5065542
  • 89175-310
    ULPA Filtered, Double Width
    ULPA Filtered, Double Width
    208/230 V, 60 Hz, (US)
    Stainless steel
    UL, ETL, CAN/CSA
    Log in for availability
    Log in for availability
    Each
    Each - $80,410.00
    $80,410.00
    Each
       $80,410.00
    La quantité minimale de commande est de 1
    Supplier Number: 5065547
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Glove Box Interior Shelves Kit, 76.2W x 15.2D cm (30 x 6")
    Glove Box Interior Shelves Kit, 76.2W x 15.2D cm (30 x 6")
    32940-080
    Log in for availability
    Each - $1,120.00
Product Information
  • Catalog #
    Availability
    Price
  • Add-A-Valve Kit, Two Manual Valves for Transfer Chamber
    Add-A-Valve Kit, Two Manual Valves for Transfer Chamber
    89194-714
    Log in for availability
    Each - $2,420.00
Product Information
  • Catalog #
    Availability
    Price
  • Add-A-Valve Kit, Two Manual Valves for Main Chamber
    Add-A-Valve Kit, Two Manual Valves for Main Chamber
    89194-716
    Log in for availability
    Each - $2,060.00
Product Information
  • Catalog #
    Availability
    Price
  • Recirculation Kit
    Recirculation Kit
    89194-712
    Log in for availability
    Each - $1,050.00
Product Information
  • Catalog #
    Availability
    Price
  • Hard Duct Connector
    Hard Duct Connector
    89194-710
    Log in for availability
    Each - $896.00
Product Information
  • Catalog #
    Availability
    Price
  • Thimble Exhaust Connector, 6", (For connection to house exhaust or remote blower)
    Thimble Exhaust Connector, 6", (For connection to house exhaust or remote blower)
    89194-708
    Log in for availability
    Each - $1,130.00
Product Information
  • Catalog #
    Availability
    Price
  • Interior Glove Port Cover
    Interior Glove Port Cover
    15202-521
    Log in for availability
    Each - $2,380.00
Product Information
  • Catalog #
    Availability
    Price
  • HEPA Inlet/Exhaust Filter
    HEPA Inlet/Exhaust Filter
    32940-085
    Log in for availability
    Each - $1,130.00
Product Information
  • Catalog #
    Availability
    Price
  • Carbon Exhaust Filter
    Carbon Exhaust Filter
    32940-090
    Log in for availability
    Each - $2,060.00
Product Information
  • Catalog #
    Availability
    Price
  • Exterior Monitor Tubing Kit (Use with Moisture Monitors 5244900 and 5244901)
    Exterior Monitor Tubing Kit (Use with Moisture Monitors 5244900 and 5244901)
    89175-442
    Log in for availability
    Each - $427.00
Product Information
  • Catalog #
    Availability
    Price
  • Add-A-Valve Installation Kit
    Add-A-Valve Installation Kit
    89194-724
    Log in for availability
    Each - $161.00
Product Information
  • Catalog #
    Availability
    Price
  • Blower Foot Switch
    Blower Foot Switch
    89194-706
    Log in for availability
    Each - $1,050.00
Product Information
  • Catalog #
    Availability
    Price
  • Positive Pressure Conversion Kit
    Positive Pressure Conversion Kit
    89194-704
    Log in for availability
    Each - $1,050.00
Product Information
  • Catalog #
    Availability
    Price
  • Thimble Exhaust Connectors, 5" (For conection to FilterMate Portable Exhauster)
    Thimble Exhaust Connectors, 5" (For conection to FilterMate Portable Exhauster)
    89200-778
    Log in for availability
    Each - $1,440.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.