Order Entry
Canada
ContactUsLinkComponent
Nuclear and Cytoplasmic Extraction Kit, G-Biosciences
Nuclear and Cytoplasmic Extraction Kit, G-Biosciences
Catalog # CA82022-600
Supplier:  G-Biosciences
CAS Number:  
undefined
Nuclear and Cytoplasmic Extraction Kit, G-Biosciences
Catalog # CA82022-600
Supplier:  G-Biosciences
Supplier Number:  786-182
CAS Number:  

Specifications

  • Description:
    Nuclear & Cytoplasmic Extraction Kit
  • Size:
    100 Preps
  • Cat. no.:
    CA82022-600
  • Application:
    Extracts Proteins, Maintains Biological Activity, Ensures Good Recovery
  • Supplier No.:
    786-182

Specifications

About this item

For the enrichment of cytoplasmic and nuclear fractions from cultured cells and tissues

This kit is based on organic buffers and contains a proprietary combination of various salts and agents. The kit is provided with reagents for solubilization of the nuclear fraction. This kit is designed for >50 preps, depending on the tissue used, where one prep is 20x10^6 mammalian cells or 100mg mammalian tissue. This kit is designed for >50 preps, depending on the tissue used, where one prep is 20x10^6 mammalian cells or 100 mg mammalian tissue.

The kit generates a cleaner separation of cytoplasmic proteins from nuclear proteins and is ideal for expression and transport studies.