Orders LinkContactUsLinkComponent
Flotation Work Stations™, Low Profile, TBS®
Flotation Work Stations™, Low Profile, TBS®
Flotation Work Stations™, Low Profile, TBS®
Certificates
Flotation Work Stations™, Low Profile, TBS®
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Equip your lab with a TBS® Flotation Bath and make your tissue preparation easier, safer and more efficient
Imagine the practicality of a removable glass dish, the long-term aesthetics afforded by a scratch and chemical resistant plastic housing, the optimum specimen viewing provided by a high contrast background illuminated by an LED light array and the dependability & safety of up-to-date microprocessor controlled electronics combined with the convenience of a HISTO/Orientator™ for flattening sections and a heated slide dryer all in one compact unit.
For those flotation bath applications not requiring the added convenience of a slide dryer or HISTO/Orientator™, the basic TBS® flotation bath provides all the functional designs of the Flotation Workstation models at an affordable price. Choose from the new 8 x 8 x 2 1/4" design or, for those labs wanting a larger work surface, there is the new 8 x 11 x 2" design.
TBS’ Flotation Workstation & Tissue Flotation Baths are now available in low-profile 8 x 8 x 2 1/4" deep dish versions, and, for those labs wanting to float larger ribbons, 8 x 11 x 2" versions are now available.
Certifications: TUV/CE/FCC certified for 89207-134, 89207-132, 89207-130, 89207-128, 89207-126, 89207-124, 89207-122, and 89207-120.
Ordering information: Flotation Work Stations™ and Tissue Flotation Baths include removable glass bowl, built-in clock & LED light array.
Specifications for Product Family
Specifications
  • Catalog No:
  • 89207-120
  • 89207-122
  • 89207-124
  • 89207-126
  • 89207-128
  • 89207-130
  • 89207-132
  • 89207-134
  • Electrical:
  • 100-120VAC
  • 220-240VAC
  • 100-120VAC
  • 220-240VAC
  • 100-120VAC
  • 220-240VAC
  • 100-120VAC
  • 220-240VAC
  • Dimensions:
  • 29.8 x 47.6 x 13.3 cm (113/4 x 183/4 x 51/4")
  • 29.8 x 47.6 x 13.3 cm (113/4 x 183/4 x 51/4")
  • 38.1 x 47.6 x 13.3 cm (15 x 183/4 x 51/4")
  • 38.1 x 47.6 x 13.3 cm (15 x 183/4 x 51/4")
  • 35.6 x 45.7 x 15.2 cm (14 x 18 x 6")
  • 35.6 x 45.7 x 15.2 cm (14 x 18 x 6")
  • 38.1 x 47.6 x 13.3 cm (15 x 183/4 x 51/4")
  • 38.1 x 47.6 x 13.3 cm (15 x 183/4 x 51/4")
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 89207-120
    Tissue Flotation Bath™, microprocessor controlled w/8" x 8" x 2 1/4" deep dish removable glass bowl, built-in clock & LED light array
    Tissue Flotation Bath™, microprocessor controlled w/8" x 8" x 2 1/4" deep dish removable glass bowl, built-in clock & LED light array
    Log in for availability
    Log in for availability
    Each
    Each - $4,840.76
    $4,840.76
    Each
       $4,840.76
    Supplier Number: TFB-S
    Specifications:
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
    More
    Specifications
    Cat. No.
    89207-120
    Electrical
    100-120VAC
    Dimensions
    29.8 x 47.6 x 13.3 cm (113/4 x 183/4 x 51/4")
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
  • 89207-122
    Tissue Flotation Bath™, microprocessor controlled w/8" x 8" x 2 1/4" deep dish removable glass bowl, built-in clock & LED light array
    Tissue Flotation Bath™, microprocessor controlled w/8" x 8" x 2 1/4" deep dish removable glass bowl, built-in clock & LED light array
    Log in for availability
    Log in for availability
    Each
    Each - $4,840.76
    $4,840.76
    Each
       $4,840.76
    Supplier Number: TFB-S-220
    Specifications:
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
    More
    Specifications
    Cat. No.
    89207-122
    Electrical
    220-240VAC
    Dimensions
    29.8 x 47.6 x 13.3 cm (113/4 x 183/4 x 51/4")
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
  • 89207-124
    Tissue Flotation Bath™, microprocessor controlled w/11" x 8" removable glass bowl, built-in clock & LED light array
    Tissue Flotation Bath™, microprocessor controlled w/11" x 8" removable glass bowl, built-in clock & LED light array
    Log in for availability
    Log in for availability
    Each
    Each - $6,280.42
    $6,280.42
    Each
       $6,280.42
    Supplier Number: TFB-L
    Specifications:
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
    More
    Specifications
    Cat. No.
    89207-124
    Electrical
    100-120VAC
    Dimensions
    38.1 x 47.6 x 13.3 cm (15 x 183/4 x 51/4")
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
  • 89207-126
    Tissue Flotation Bath™, microprocessor controlled w/11" x 8" removable glass bowl, built-in clock & LED light array
    Tissue Flotation Bath™, microprocessor controlled w/11" x 8" removable glass bowl, built-in clock & LED light array
    Log in for availability
    Log in for availability
    Each
    Each - $5,225.86
    $5,225.86
    Each
       $5,225.86
    Supplier Number: TFB-L-220
    Specifications:
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
    More
    Specifications
    Cat. No.
    89207-126
    Electrical
    220-240VAC
    Dimensions
    38.1 x 47.6 x 13.3 cm (15 x 183/4 x 51/4")
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
  • 89207-128
    Flotation Work Station™, microprocessor controlled w/removable glass bowl 8" x 8", slide dryer 2" x 5" and tissue orientator 2" x2"
    Flotation Work Station™, microprocessor controlled w/removable glass bowl 8" x 8", slide dryer 2" x 5" and tissue orientator 2" x2"
    Log in for availability
    Log in for availability
    Each
    Each - $6,533.78
    $6,533.78
    Each
       $6,533.78
    Supplier Number: FWS-S
    Specifications:
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
    More
    Specifications
    Cat. No.
    89207-128
    Electrical
    100-120VAC
    Dimensions
    35.6 x 45.7 x 15.2 cm (14 x 18 x 6")
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
  • 89207-130
    Flotation Work Station™, microprocessor controlled w/removable glass bowl 8" x 8", slide dryer 2" x 5" and tissue orientator 2" x2"
    Flotation Work Station™, microprocessor controlled w/removable glass bowl 8" x 8", slide dryer 2" x 5" and tissue orientator 2" x2"
    Log in for availability
    Log in for availability
    Each
    Each - $6,261.23
    $6,261.23
    Each
       $6,261.23
    Supplier Number: FWS-S-220
    Specifications:
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
    More
    Specifications
    Cat. No.
    89207-130
    Electrical
    220-240VAC
    Dimensions
    35.6 x 45.7 x 15.2 cm (14 x 18 x 6")
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
  • 89207-132
    Flotation Work Station™, microprocessor controlled w/11" x 8" removable glass bowl w/built-in LED light array, 2" x 8" slide dryer and 2" x 2" HISTO/Orientator™
    Flotation Work Station™, microprocessor controlled w/11" x 8" removable glass bowl w/built-in LED light array, 2" x 8" slide dryer and 2" x 2" HISTO/Orientator™
    Log in for availability
    Log in for availability
    Each
    Each - $7,116.94
    $7,116.94
    Each
       $7,116.94
    Supplier Number: FWS-L
    Specifications:
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
    More
    Specifications
    Cat. No.
    89207-132
    Electrical
    100-120VAC
    Dimensions
    38.1 x 47.6 x 13.3 cm (15 x 183/4 x 51/4")
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
  • 89207-134
    Flotation Work Station™, microprocessor controlled w/11" x 8" removable glass bowl w/built-in LED light array, 2" x 8" slide dryer and 2" x 2" HISTO/Orientator™
    Flotation Work Station™, microprocessor controlled w/11" x 8" removable glass bowl w/built-in LED light array, 2" x 8" slide dryer and 2" x 2" HISTO/Orientator™
    Log in for availability
    Log in for availability
    Each
    Each - $7,347.12
    $7,347.12
    Each
       $7,347.12
    Supplier Number: FWS-L-220
    Specifications:
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
    More
    Specifications
    Cat. No.
    89207-134
    Electrical
    220-240VAC
    Dimensions
    38.1 x 47.6 x 13.3 cm (15 x 183/4 x 51/4")
    cUL Listed
    CSA Certified
    Small Business Enterprise
    UL Listed
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Replacement Bowl, for models 8x8 models only.
    Replacement Bowl, for models 8x8 models only.
    CA27900-130
    Log in for availability
    Each - $173.23
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)