ContactUsLinkComponent
Glycoprotein Staining Kit, G-Biosciences
Glycoprotein Staining Kit, G-Biosciences
Catalog # CA82023-286
Supplier:  G-Biosciences
CAS Number:  
undefined
Glycoprotein Staining Kit, G-Biosciences
Catalog # CA82023-286
Supplier:  G-Biosciences
Supplier Number:  786-254
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Glycoprotein Staining Kit
  • Size:
    10 mini gels
  • Cat. no.:
    CA82023-286
  • Supplier No.:
    786-254
Specifications
About this item
G-Biosciences offers the Glycoprotein Staining kit for the highly sensitive detection of glycoproteins following gel electrophoresis or protein transfer to nitrocellulose membranes
G-Biosciences' Glycoprotein Staining kit uses an enhanced Periodic Acid-Schiff (PAS) method for detection of glycoprotein sugars. The supplied oxidizing agent first oxidizes the cis-diol sugar groups to aldehydes. The aldehyde groups react with the sensitive Glyco-Stain Solution forming Schiff bonds and producing strong magenta color bands.
G-Biosciences' Glycoprotein Staining kit is supplied with an enhanced Coomassie stain, called RAPIDstain™, that enhances the glycoprotein staining. RAPIDstain™ can also be used after glycoprotein staining to detect non-glycosylated proteins. The kit is sufficient for 10 mini gels (8 x 8 cm) or 20 nitrocellulose membrane (8 x 8 cm).
The Glycoprotein Staining kit is highly convenient as all the key reagents required for staining are supplied and a unique positive & negative control is included. In addition, the kit allows for the detection of glycosylated and non-glycosylated proteins on a single gel or membrane.