Order Entry
Canada
ContactUsLinkComponent
BLOT-QuickBlocker™ Modified Milk Protein Blocking Agent, G-Biosciences
BLOT-QuickBlocker™ Modified Milk Protein Blocking Agent, G-Biosciences
Catalog # CA82021-234
Supplier:  G-Biosciences
CAS Number:  
BLOT-QuickBlocker™ Modified Milk Protein Blocking Agent, G-Biosciences
Catalog # CA82021-234
Supplier:  G-Biosciences
Supplier Number:  786-011
CAS Number:  

Specifications

  • Description:
    BLOT-QuickBlocker™
  • Size:
    175 g
  • Cat. no.:
    CA82021-234
  • Application:
    Modified Milk Protein Blocker for Immunodetection Assays
  • Supplier No.:
    786-011

Specifications

About this item

G-Biosciences' BLOT-QuickBlocker™ is a novel modified milk protein that is highly soluble and does not inhibit peroxidase detection

G-Biosciences' BLOT-QuickBlocker™ is readily soluble and produces a semi-clear solution. With a blocking time of about 30 to 60 minutes, this blocking agent is suitable for immunodetection assays such as Western blots and dot blots.

The modified milk protein has high blocking efficiency with a clear background.

Ordering information: Accessories available.