Nalgene® Narrow-Mouth Packaging Bottles, Translucent Amber HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These translucent amber high density polyethylene bottles are leakproof.
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
They include opaque amber polypropylene closures. These bottles have excellent chemical resistance and have a robust construction for packaging use, and are designed for storage of light sensitive liquids.
Ordering information: They are bulk packed for packaging applications.
Customers Who Bought This Also Bought