Order Entry
Canada
ContactUsLinkComponent
Pierce™ Substrate for Western Blotting, ECL Plus, Thermo Scientific
Pierce™ Substrate for Western Blotting, ECL Plus, Thermo Scientific
Catalog # CAPI80196
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Substrate for Western Blotting, ECL Plus, Thermo Scientific
Catalog # CAPI80196
Supplier:  Thermo Scientific
Supplier Number:  80196
CAS Number:  

Specifications

  • Description:
    Pierce™ ECL plus western blotting substrate, for 10 mini-gel blots
  • Size:
    100 ml (32/5 oz.)
  • Cat. no.:
    CAPI80196
  • For Membrane Area:
    1000 cm²
  • Supplier No.:
    80196

Specifications

About this item

The Pierce™ ECL Plus Western Blotting Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for Western blot detection using X-ray film or CCD- or laser-based imagers.

  • Easy to use, high quality and performance
  • Higher sensitivity - detect targets down to the low-picogram level
  • Longer signal duration - sustained light output for as long as 5 hours
  • More imaging options - X-ray, CCD or laser-based imagers

Pierce™ ECL 2 Substrate generates acridinium esters when it reacts with HRP

Pierce™ ECL 2 substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. A broad dynamic range and excellent sensitivity combined with a long-lasting signal make Pierce ECL 2 Substrate a desirable Western blotting substrate.

As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers.

Delivery: Kit includes proprietary acridan and peroxide solutions.