Immuno Standard High-binding Modules, Thermo Scientific
Catalog # CA62407-137
Supplier: Thermo Fisher Scientific
Specifications
- Volume:400 µl
- Well Volume:350 µL
- Cat. no.:CA62407-137
- Surface Treatment:MaxiSorp™
- Supplier No.:434797
Specifications
About this item
Immuno Standard Modules are high binding and are developed for ELISA techniques and fit standard equipment
Their format provides flexible use of components with small sample sizes. The modules offer lot-to-lot reliability, well-to-well reproducibility and are backed by industry leading quality standards and certifications.
Certifications: CSA Certified. Products specified in chart are certified for Adsorption of IgG; CV of <5% between wells; all results ±10% from mean; and background of all wells ±0.005 absorbance units from mean.