ContactUsLinkComponent
SDS Detection & Estimation Reagent Kit, G-Biosciences
SDS Detection & Estimation Reagent Kit, G-Biosciences
Catalog # CA82021-290
Supplier:  G-Biosciences
CAS Number:  
SDS Detection & Estimation Reagent Kit, G-Biosciences
Catalog # CA82021-290
Supplier:  G-Biosciences
Supplier Number:  786-129
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    SDS-Detection Kit
  • Cat. no.:
    CA82021-290
  • Supplier No.:
    786-129
Specifications
About this item
A reagent kit for detection and estimation of SDS in a sample
The working range of the assay is 0.02-0.2% SDS.
Mix the test sample in the extraction buffer reagents provided with the kit. If SDS is present in the sample, a blue color is extracted that can be quantitatively measured.
Ordering information: Accessories not included.