Order Entry
Canada
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

1 g
Catalog # CA95057-556
Supplier:   G-Biosciences
Supplier #:  786-262
Trypsin
Catalog # CA95057-556
Supplier:  G-Biosciences
CAS Number:  
CAS Number:  
Price
$518.40
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Trypsin is a serine endopeptidase that specifically cleaves peptide bonds on the carboxy side of s-aminoethyl cysteine, arginine and lysine residues

This trypsin is purified from bovine pancreas, 1X crystallized, dialyzed against 1mM HCl and lyophilized. >150 TAME untis/ mg protein. One Unit hydrolyzes 1µmole of p-toluene-sulfonyl-L-arginine methyl ester (TAME) per minute at 25°C, pH 8.2, in the presence of 10mM calcium.

For mass spectrometry sequence analysis, we recommend our ultra-pure MSG-Trypsin™ that is specifically designed for mass spectrometry.

Typically there is little or no cleavage at arginyl-proline and lysyl-proline bonds.

Ordering information: Accessories not included.

Documents

Specifications

Specifications

  • Size:
    1 g
  • Cat. No.:
    CA95057-556
  • Form:
    Lyophilized
  • Enzyme name:
    Trypsin

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.