All Hei-VAP models come with a detachable operating panel with 4' (ft) tethered cable that provides unparalleled safety by attachment outside of fume hoods and greater visual control
Available in Hand Lift (HL) configuration. Two large dial controls for adjustment of rotation speed and bath temperature. Digital models come with a digital display for set and actual heating bath temperature. Optional Manual Vacuum Controller for easy distillations.
Hei-VAP Value “The Collegiate”– Hand lift for all basic applications.
Hei-VAP rotary evaporators have a stainless steel heat bath with 2 independent safety circuits for added safety that can accommodate up to a 5 liter flask and has a maximum bath temperature of 210°C. Our non-sticking vapor tube-clamping sleeve eliminates hassle and provides operator safety during removal of vapor tube. Heidolph®’s long lasting vacuum seal can reduce your consumable costs and makes the Hei-VAP your clear choice for rotary evaporation. All models can be packaged with Heidolph® vacuum pumps, chillers, and other accessories to make your purchasing solutions simple.