Specifications
- Description:9 x 51 mm ISCI dish
- Cat. no.:CA89106-782
- Surface Treatment:Untreated
- Supplier No.:150265
Specifications
About this item
Thermo Scientific Nunc® provide an extensive portfolio of products intended for use in human In Vitro Fertilization (IVF)
The Nunc® IVF ICSI Dish maintains an optimal temperature with true flat bottom, as well as increased safety with the easy grip design. The low profile provides optimal angles during ICSI process, while the super polished optical center allows clear viewing of the sample.
Thermo Scientific Nunc® InVitro Fertilization Products provide security, safety and reproducibility for your valuable samples, and are uniquely certified based on a 1-cell stage mouse embryo toxicity test. Stringent lot-level manufacturing controls ensure viability, reliability and reproducibility. Lid is Present.
Products are produced under ISO 13485, MEA tested, CE marked class II medical device and FDA cleared for use in IVF.