Order Entry
Canada
ContactUsLinkComponent
 
Isopropylbenzene (Cumene), Standard, 2000 μg/ml, Restek
  RK30294
 :  Restek
 :  
undefined
Isopropylbenzene (Cumene), Standard, 2000 μg/ml, Restek
  RK30294
 :  Restek
 :  30294
 :  

 

  • Pack type:
    Ampule
  • CAS No.:
    98-82-8
  • Size:
    1 ml
  • Formula:
    C₉H₁₂
  • Description:
    Isopropylbenzene (Cumene)
  • Cat. no.:
    RK30294
  • Supplier No.:
    30294

 

 

Solvent packaged in purge-and-trap-grade methanol.

  • 2000 μg/mL in P&T methanol, 1 mL/ampoule

Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.