Ethanol, Standard, 2000 μg/ml, Restek
Catalog # RK30288
Supplier: Restek
CAS Number:
Specifications
- Pack type:Ampule
- CAS No.:64-17-5
- Size:1 ml
- Formula:C₂H₆O
- Description:Ethanol
- Cat. no.:RK30288
- Supplier No.:30288
Specifications
About this item
Solvent packaged in purge-and-trap-grade methanol.
- 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.