Order Entry
Canada
ContactUsLinkComponent
Chloroethane, Standard, 2000 μg/ml, Restek
Chloroethane, Standard, 2000 μg/ml, Restek
Catalog # RK30263
Supplier:  Restek
CAS Number:  
undefined
Chloroethane, Standard, 2000 μg/ml, Restek
Catalog # RK30263
Supplier:  Restek
Supplier Number:  30263
CAS Number:  

Specifications

  • Pack type:
    Ampule
  • CAS No.:
    75-00-3
  • Size:
    1 ml
  • Formula:
    C₂H₅Cl
  • Description:
    Chloroethane
  • Cat. no.:
    RK30263
  • Supplier No.:
    30263

Specifications

About this item

Solvent packaged in purge-and-trap-grade methanol.

  • 2000 μg/mL in P&T methanol, 1 mL/ampoule

Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.