Carbon disulfide, Standard, 2000 μg/ml, Restek
RK30258
: Restek
:
- Pack type:Ampule
- CAS No.:75-15-0
- Size:1 ml
- Formula:CS₂
- Description:Carbon disulfide
- Cat. no.:RK30258
- Supplier No.:30258
Solvent packaged in purge-and-trap-grade methanol.
- 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.