Orders LinkContactUsLinkComponent
Semi-quantitative test strips QUANTOFIX Nitrite
100 Sticks
Catalog # 60787-734
Supplier:   Macherey-Nagel
 :  91311
Semi-quantitative test strips QUANTOFIX Nitrite
Catalog # 60787-734
Supplier:  Macherey-Nagel
CAS Number:  
Semi-quantitative test strips QUANTOFIX Nitrite
Semi-quantitative test strips QUANTOFIX Nitrite
100 Sticks
CAS Number:  
Price
$141.54
Log in for availability
Pack of 100
Product Details & Documents
For semi-quantitative determinations
Gradation: 0 to 80mg/L. Color changes from white to red/violet.
Specifications
Specifications
  • Pack type:
    100 Sticks
Specifications