Anti-6xHis Rabbit Polyclonal Antibody (DyLight® 800)
Catalog # CA600-445-382
Supplier: Rockland Immunochemical
Specifications
- Antibody type:Tag
- Antigen name:6X HIS Affinity Tag
- Antigen symbol:6xHis
- Clonality:Polyclonal
- Conjugation:DyLight® 800
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Size:100 µg
- Antigen synonyms:Histidine Epitope Tag|His Tag|Histidine Tag
- Cat. no.:CA600-445-382
- Supplier No.:600-445-382
Specifications
About this item
Primary Rabbit Anti-6X HIS EPITOPE TAG Reacts with Pan
Type: Tag
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan