Anti-GST Mouse Polyclonal Antibody [clone: 3D4] (DyLight® 649)
Catalog # CA200-343-200
Supplier: Rockland Immunochemical
Specifications
- Antibody type:Primary
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Polyclonal
- Conjugation:DyLight® 649
- ELISA:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Multiple
- Western blot:Yes
- Size:100 µg
- ChIP:Yes
- Neutralisation:Yes
- Antigen synonyms:GSTP1-1|PI|DFN7|GST3|GSTP1|FAEES3
- Cat. no.:CA200-343-200
- Supplier No.:200-343-200
Specifications
About this item
Primary Mouse Anti-Glutathione-S-Transferase (GST) Reacts with Pan
Type: Primary
Antigen: Glutathione-S-Transferase
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope:
Host: Mouse
Isotype:
Reactivity: Pan