Anti-MYC Rabbit Polyclonal Antibody (DyLight® 549)
Catalog # CA600-442-381
Supplier: Rockland Immunochemical
Specifications
- Antibody type:Tag
- Antigen name:V-myc Avian Myelocytomatosis Viral Oncogene homolog
- Antigen symbol:MYC
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Size:100 µg
- Antigen synonyms:bHLHe39|Myc2|Nird|AU016757|Niard
- Cat. no.:CA600-442-381
- Supplier No.:600-442-381
Specifications
About this item
Primary Rabbit Anti-MYC EPITOPE TAG Reacts with Pan
Type: Tag
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan