Order Entry
Canada
ContactUsLinkComponent
Anti-MMP9 Rabbit Polyclonal Antibody
Anti-MMP9 Rabbit Polyclonal Antibody
Catalog # CA80053-088
Supplier:  MilliporeSigma
Anti-MMP9 Rabbit Polyclonal Antibody
Catalog # CA80053-088
Supplier:  MilliporeSigma
Supplier Number:  444236-500UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 9
  • Antigen symbol:
    MMP9
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Marmoset,
    Human
  • Western blot:
    Yes
  • Size:
    500 µL
  • Antigen synonyms:
    92kDa type IV collagenase)|MMP-9|GELB|92 kDa gelatinase|MANDP2|matrix metalloproteinase 9 (gelatinase B|92 kDa type IV collagenase|type V collagenase|matrix metalloproteinase-9|92kDa gelatinase|macrophage gelatinase|CLG4B
  • Cat. no.:
    CA80053-088
  • Supplier No.:
    444236-500UL

Specifications

About this item

Primary Rabbit Anti-MMP-9 Reacts with Human, Marmoset

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Marmoset