Orders LinkContactUsLinkComponent
VWR® TBS Buffer, 20X Liquid
4 L
  97064-338
 :   VWR
 :  J640-4L
VWR® TBS Buffer, 20X Liquid
  97064-338
 :  VWR
 :  
VWR® TBS Buffer, 20X Liquid
VWR® TBS Buffer, 20X Liquid
4 L
 :  
$480.65
Each (4l)
A 1X solution of Tris Buffered Saline (TBS) contains 140 mM Sodium Chloride, 3.0 mM Potassium Chloride and 25 mM Tris.
  • Size:
    4 L