Transfection Reagent Diluent, 150 mM NaCl Solution, Polyplus-transfection
Catalog # CA89129-970
Supplier: Polyplus Transfection
CAS Number:
Specifications
- DNA:Yes
- Description:Transfection reagent diluent, 150 mM NaCl solution
- Cat. no.:CA89129-970
- Buffer Volume:50 ml
- Supplier No.:702-50
Specifications
About this item
This 150 mM NaCl solution is recommended as dilution reagent for DNA transfection with jetPEI®.