Order Entry
Canada
ContactUsLinkComponent
PULSin®, protein delivery reagent, Polyplus-transfection
PULSin®, protein delivery reagent, Polyplus-transfection
Catalog # CA89129-956
CAS Number:  
undefined
PULSin®, protein delivery reagent, Polyplus-transfection
Catalog # CA89129-956
Supplier Number:  101000010
CAS Number:  

Specifications

  • Protein delivery:
    Yes
  • Description:
    PULSin®, protein delivery reagent
  • Size:
    400 µL
  • Cat. no.:
    CA89129-956
  • Buffer Volume:
    20 ml
  • No. of Reactions:
    24 (6 wells)
  • Supplier No.:
    501-04

Specifications

About this item

PULSin® protein delivery reagent is dedicated to the delivery of proteins, antibodies and peptides into cells and offers a powerful approach to functional studies

  • Effective and robust delivery into the cytoplasm
  • Delivery to a wide variety of cells including primary cells and Hard-to-transfect cells
  • Highly efficient delivery
  • Easy to use

PULSin® can be used to target intracellular proteins with antibodies in living cells without fixation.

Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Every batch of PULSin™ is tested by delivering R-phycoerythrin into HeLa cells.
Delivery: Supplied with 20 µg R-Phycoerythrin (positive control) and Hepes dilution buffer. 0.4 ml of PULSin® protein delivery reagent is sufficient for 24 reactions in 6-well plates.