Nunc® Centrifuge Tubes, Polypropylene, Conical, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Nunc® conical sterile polypropylene centrifuge tubes are premium, high quality, and environmentally friendly conical tubes.
- Plastic labware is a safer alternative to glass without sacrificing accuracy
These tubes have a high RCF rating that enables a greater range of applications, from low speed to super speed centrifugation. They are also USP Class VI, non-pyrogenic, non-cytotoxic and RNase/DNase free to ensure the highest performance.
Thermo Scientific Nunc® labware products are made from high purity resins, and molded using our state-of-the-art processes.
The tubes have leakproof construction and offer increased traceablilty with the largest writing area on the market. Additionally, they offer a reduced contamination risk with a recyclable plastic rack.
Ordering information: Tubes 89174-470 and 89174-474 includes reusable and recyclable polypropylene rack.
Related Information
Certificates
Search by Lot Number89174-468_J7AF617116 89174-468_JAAF9Z7116 89174-470_J7AF617117 89174-472_J7AF618118 CA89174-470_J4AF317117
Conformity declaration
More
89174-470
Images 360
Thermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Customers Who Bought This Also Bought