Specifications
- Description:Kidney Stone Collection Cup
- Cat. no.:89170-924
- Supplier No.:75109
Specifications
About this item
Designed to capture renal calculi and intended for use with patients attempting to pass stones following extracorporeal shock wave lithotripsy
Designed to capture renal calculi and intended for use with patients attempting to pass stones following extracorporeal shock wave lithotripsy
The urine-straining device is a collapsible plastic strainer cup that can be used by both men and women and includes a nylon screen at the bottom of the cup for stone capture. Once the stone is captured, it can be placed in the storage space under the lid and saved for analysis.