Specifications
- Description:ICON® 25 hCG Pregnancy Test
- Cat. no.:CA63900-010
- Supplier No.:43025A
Specifications
About this item
Highly sensitive, one-step immunochemical rapid test for qualitative detection of human chorionic gonadotropin in urine or serum to aid in early pregnancy detection.
- Sensitive (25 mIU/mL)
- Specificity > 99%
- One-step (test time 3 minutes for urine, 5 minutes for serum)
- Easy to use
Meets the requirements of physicians and hospitals for ease-of-use, value, and accuracy.