DrySlide™ Indole Slide, BD Biosciences
Catalog # CA90002-574
Supplier: BD
Specifications
- Description:Indole Slide
- Cat. no.:CA90002-574
- Supplier No.:231748
Specifications
About this item
Disposable slide employs a dry reagent cassette impregnated with 5% p-dimethylaminobensaldehyde (DMABA) for the rapid spot-identification of indole producing bacteria from primary cultures
Divided into four large segments to enable multiple tests on one slide. Provides results in just 30 seconds