About this item
BD filter needles feature a molded-in 5-micron filter membrane that reduces larger particles during fluid aspiration, reconstitution or transfer, and during medication withdraw from glass reservoirs.
- Reduce glass particles when preparing medications
The Nokor point - The vented point design features a cannula side port for you to place fluid on the container side wall during reconstitution, helping reduce foaming or spray.
Reduced internal pressure - Vented needles with a side port reduce internal container pressure.