About this item
Blunt needles require a higher puncture penetration force than a conventional sharp, so they are less likely to cause needlestick injuries while preparing and filling medications from vials, ampoules, and IV bags.
These 18-gauge needles allow for rapid transfer and aspiration without coring and are long enough to easily penetrate single and double-septum barriers. Needles feature a standard Luer hub for standardization and are available already attached to syringes for ready use in medication preparation. The red hub and shield are easily identified as a unique needle.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot NumberBD305180_9058677 BD305180_9269053 BD305180_9269059 BD305211_9113719 CABD305180_8331645