ContactUsLinkComponent
INTAKE™ Reagent Filter, POREX®
INTAKE™ Reagent Filter, POREX®
Catalog # 28295-546
Supplier:  Porex
undefined
INTAKE™ Reagent Filter, POREX®
Catalog # 28295-546
Supplier:  Porex
Supplier Number:  6603
Specifications
  • Description:
    Reagent Filter
  • Cat. no.:
    28295-546
  • Supplier No.:
    6603
Specifications
About this item
This disposable filter is made of ultra-high molecular weight polyethylene (UHMWPE), and can filter all particulate matter down to 10 µm.
The filter is for use with automated instruments, benchtop dispensers, or anywhere an agent is drawn from a master storage bottle. Attaches to equipment line with glass weight (not provided).
Ordering information: Designed for use with glass or plastic collection tubes.