Orders LinkContactUsLinkComponent
Suite of Columns, Method Development Kits, MicroSolv
Suite of Columns, Method Development Kits, MicroSolv
Certificates
Suite of Columns, Method Development Kits, MicroSolv
Suite of Columns, Method Development Kits, MicroSolv
11 Options Available Below
Product Details & Documents
Complete set of columns for method developers that need and want the full range of selectivities on hand at all times.
Each set contains one each of Bidentate C18™, UDC-Cholesterol™, Bidentate C8™, Diamond-Hydride™ and Silica-C™ columns in the same configuration.
Range from extremely hydrophobic to extremely hydrophilic, from low pH to high pH, from reversed phase to aqueous normal phase to normal phase with only five columns.
 : Guard column suites contain five guard columns with or without a holder.
Product Family Options
Columns
Product Information
  • Column Format
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • 97035-104
    5 Column Suite
    5 Column Suite
    Capillary
    100 Å
    4 µm
    1.00 mm
    150 mm
    Log in for availability
    Log in for availability
    Each
    Each - $10,250.52
    $10,250.52
    Each
       $10,250.52
    Supplier Number: 40100-15P-1
  • 97035-098
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    2.10 mm
    50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $7,747.95
    $7,747.95
    Each
       $7,747.95
    Supplier Number: 40100-05P-2
  • 97035-106
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    2.10 mm
    75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $8,187.85
    $8,187.85
    Each
       $8,187.85
    Supplier Number: 40100-7.5P-2
  • 97035-068
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    2.10 mm
    100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $9,465.71
    $9,465.71
    Each
       $9,465.71
    Supplier Number: 40100-10P-2
  • 97035-062
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    2.10 mm
    150 mm
    Log in for availability
    Log in for availability
    Each
    Each - $10,609.76
    $10,609.76
    Each
       $10,609.76
    Supplier Number: 40100-15P-2
  • 97035-100
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    3.00 mm
    50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $7,747.95
    $7,747.95
    Each
       $7,747.95
    Supplier Number: 40100-05P-3
  • 97035-058
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    4.60 mm
    50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $7,980.45
    $7,980.45
    Each
       $7,980.45
    Supplier Number: 40100-05P
  • 97035-066
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    4.60 mm
    75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $8,433.52
    $8,433.52
    Each
       $8,433.52
    Supplier Number: 40100-7.5P
  • 97035-102
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    4.60 mm
    100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $9,235.10
    $9,235.10
    Each
       $9,235.10
    Supplier Number: 40100-10P
  • 97035-060
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    4.60 mm
    150 mm
    Log in for availability
    Log in for availability
    Each
    Each - $10,558.00
    $10,558.00
    Each
       $10,558.00
    Supplier Number: 40100-15P
  • 97035-064
    5 Column Suite
    5 Column Suite
    Analytical
    100 Å
    4 µm
    4.60 mm
    250 mm
    Log in for availability
    Log in for availability
    Each
    Each - $11,233.48
    $11,233.48
    Each
       $11,233.48
    Supplier Number: 40100-25P