

Acetone ≥99.5%, Finyte® for the electronics industry, J.T.Baker®
CAJT5018-20
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:PE drum, black
- Size:330 lbs.
- Cat. no.:CAJT5018-20
- Supplier No.:5018-20
- Assay (by GC):≥99.5 %
- Color (APHA):≤10
- Acidity (µeq/g):≤0.3
- Aldehydes (as HCHO):≤20000 ppb
- Alkalinity (µeq/g):≤0.5
- Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):≤0.05 %
- Methanol (CH₃OH)(by GC):≤0.05 %
- Residue after Evaporation:≤3000.0 ppb
- Water (H₂O):≤0.50 %
- Solubility in H₂O:Passes Test
- Chloride (Cl):≤200.0 ppb
- Phosphate (PO₄):≤50.0 ppb
- Trace Impurities - Aluminum (Al):≤10.0 ppb
- Trace Impurities - Antimony (Sb):≤5.0 ppb
- Trace Impurities - Arsenic (As):≤5.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤10.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤10.0 ppb
- Trace Impurities - Chromium (Cr):≤10.0 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤10.0 ppb
- Trace Impurities - Iron (Fe):≤10.0 ppb
- Trace Impurities - Lead (Pb):≤10.0 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤10.0 ppb
- Trace Impurities - Silicon:≤10.0 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤10.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤10.0 ppb
- Trace Impurities - Titanium (Ti):≤10.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤10.0 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- Particle Count at point of fill - 0.5 µm and greater:≤150 par/ml
- Particle Count at point of fill - 1.0 µm and greater:≤25 par/ml
:
:58.08 g/mol
:13,00067
:56.2 °C (1013 hPa)
:–95.4 °C
:0.792 g/cm³ (20 °C)
:<–20 °C (closed cup)
:Ambient
:200-662-2
:1090
:3,II
:Management of Change (MOC) category = R
:01-2119471330-49
:GHS02
:GHS07
:67-64-1
:CH₃COCH₃
:MFCD00008765