1,6-Dichlorohexane 98%
Product Details & Documents
Purity: 98% CAS No.: 2163-00-0 Molecular Formula: Cl(CH2)6Cl Formula Weight: 155.07 Flash Point: 73°(163°F) Refractive Index: 1.4568 Melting Point: -13° Boiling Point: 204-205° Beilstein Reference: 773673 UN No.: 3082 MDL No.: MFCD00001020
- CAS Number:2163-00-0
- Formula:ClH₂C(CH₂)₄CH₂Cl
- MDL Number:MFCD00001020
- Molecular Weight:155.07 g/mol
- Boiling Pt:204…205 °C
- Melting Pt:–13 °C
- Density:1.067
- Flash Pt:73 °C (163 °F)
- Storage Temperature:Ambient
- EINECS:218-491-7
- UN:3082
- ADR:9,III
Documents
Specifications
Specifications
- Pack type:Bottle
- Size:500 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



