Order Entry
Canada
ContactUsLinkComponent
4-Chloro-m-phenylenediamine 97%
Irritant
Health
4-Chloro-m-phenylenediamine 97%
Catalog # CAAAA15430-06
CAS Number:  
Irritant
Health
4-Chloro-m-phenylenediamine 97%
Catalog # CAAAA15430-06
Supplier Number:  A15430-06
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    5 g
  • Cat. no.:
    CAAAA15430-06
  • Supplier No.:
    A15430-06

Specifications

About this item

 :
 :142.59 g/mol
Boiling Pt:280 °C (1013 hPa)
Melting Pt:87…90 °C
EINECS:225-877-9
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:5131-60-2
Formula:C₆H₇ClN₂
MDL Number:MFCD00025284