1,1,2,2-Tetrabromoethane 97%
About this item
Purity: 97% CAS No.: 79-27-6 Molecular Formula: Br2CHCHBr2 Formula Weight: 345.67 Refractive Index: 1.6370 Boiling Point: 118-120°/15mm Merck Reference: 14,9185 UN No.: 2504 MDL No.: MFCD00000133
- CAS Number:79-27-6
- Formula:Br₂HCCHBr₂
- MDL Number:MFCD00000133
- Molecular Weight:345.65 g/mol
- Merck Index:14,9185
- Boiling Pt:118…120 °C/15 mm
- Melting Pt:–1 °C
- Density:2.967
- Storage Temperature:Ambient
- EINECS:201-191-5
- UN:2504
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 79-27-6 Molecular Formula: Br2CHCHBr2 Formula Weight: 345.67 Refractive Index: 1.6370 Boiling Point: 118-120°/15mm Merck Reference: 14,9185 UN No.: 2504 MDL No.: MFCD00000133
- CAS Number:79-27-6
- Formula:Br₂HCCHBr₂
- MDL Number:MFCD00000133
- Molecular Weight:345.65 g/mol
- Merck Index:14,9185
- Boiling Pt:118…120 °C/15 mm
- Melting Pt:–1 °C
- Density:2.967
- Storage Temperature:Ambient
- EINECS:201-191-5
- UN:2504
- ADR:6.1,III
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



