You searched for: Microscopy and staining reagents
VWR offers a broad line of reagent droppers, diagnostic kits, and antigens and antisera, for aid in identifying a variety of microscopic organisms. The visualization of bacteria by means of staining procedures is an important tool for any microbiologist to have at their disposal, and these carefully standardized and reproducible stain reagents help to ensure accurate and easily interpretable results. Stains are also available to provide early detection of positive blood cultures by fluorescence procedures.
VWR® Xylene Substitute, Lab-Grade for Histology
Supplier: VWR International
Expand 1 Items
VWR® Premium Frozen Section Compound
Supplier: VWR International
Frozen section compound is a fast-acting, water-soluble embedding compound designed for use in frozen sectioning.
Expand 1 Items
Decalcifying Solution, VWR Chemicals BDH®
Supplier: VWR International
Made with formic acid.
Expand 1 Items
Albumin-Glycerol Fixative, Azer Scientific
Supplier: VWR International
Fixative is used to adhere tissue specimens to glass slides in the histology laboratory.
Expand 2 Items
Technovit® 7100 and 8100 Embedding Kits, Electron Microscopy Sciences
Supplier: VWR International
Technovit® Glycol Methacrylate embedding kits developed by Kulzer in Germany are the most convenient to use and offer the best embedding results with improved morphology and 1 micron serial sectioning.
Expand 2 Items
VWR® HistoChoice® Clearing Agent
Supplier: VWR
Less hazardous replacement for xylene and toluene. Compatible with organic mounting media, clearing agent is designed to work with isopropanol in automated processors. It also offers faster wax dissolution and slower evaporation than xylene with no oily residue. Clearing agent can be recycled by distillation.
Expand 2 Items
Neo-Clear® Xylene Substitute, HARLECO®, EMD Millipore
Supplier: MilliporeSigma
Solvent used as a medium for clearing and deparafinizing steps in staining and tissue processing
Expand 1 Items
Entellan New® Rapid Mounting Medium, MilliporeSigma
Supplier: MilliporeSigma
Entellan New® rapid mounting medium for microscopy.
Expand 1 Items
M-GLAS™ Liquid Cover Glass, MilliporeSigma
Supplier: MilliporeSigma
M-GLAS® should be used instead of a cover glass for the homogeneous mounting of cytological specimens
Expand 1 Items
HARLECO® Bouin Fixation Fluid, Sigma-Aldrich®, MilliporeSigma
Supplier: MilliporeSigma
HARLECO® Tissue Fixative. Excellent for preserving delicate cytoplasmic structures such as cilia, mitotic spindles and astral radiations
Expand 1 Items
Entellan® Rapid Mounting Medium, MilliporeSigma
Supplier: MilliporeSigma
Entellan® rapid mounting medium for microscopy.
Expand 1 Items
Immersion Oil, MilliporeSigma
Supplier: MilliporeSigma
Immersion media are used in conjunction with immersion objectivs and are located between the surface of the specimen and the lens of the microscope
Expand 1 Items
OSTEOMOLL® Rapid Decalcifier-Solution for Histology, EMD Millipore
Supplier: MilliporeSigma
Decalcifiers for clinical diagnostic purposes and in laboratory accreditation processes.
Expand 1 Items
Histosec® Pastilles, Merck Millipore
Supplier: MilliporeSigma
Histosec® pastilles is an embedding medium for histology of samples of human origin and is a paraffin enriched with polymers.
Expand 2 Items
HARLECO® Krystalon™ Mounting Media, MilliporeSigma
Supplier: MilliporeSigma
Toulene-based
Expand 1 Items
Neo-Clear® Xylene Substitute, EMD Millipore
Supplier: MilliporeSigma
Neo-Clear® is a non hazardous alternative to xylene. It is virtually odourless, compatible with most high purity solvents and is characterised by lower evaporation rates than xylene, making it a lower health risk.
Expand 2 Items
SAF Fixative, EMD MilliporeSigma
Supplier: MilliporeSigma
HARLECO®, Acetate Buffered Formalin with Triton
Expand 2 Items
Histosec®
Supplier: MilliporeSigma
Histosec® pastilles is an embedding medium for histology of samples of human origin and is a paraffin enriched with polymers.
Expand 2 Items
Antifade Mounting Medium (Hardset), Enzo Life Sciences
Supplier: Enzo Life Sciences
Mitochondria dye.
Expand 1 Items
Antifade Mounting Medium With Dapi (Hardset), Enzo Life Sciences
Supplier: Enzo Life Sciences
Mitochondria dye.
Expand 1 Items
Bouin's Fixative
Supplier: Ricca Chemical
Picric acid-formalin-acetic acid mixture.
Expand 3 Items
Saccomanno Collection Fluid, Fixative and Preservative for Sputum Samples
Supplier: Ricca Chemical
Fixative and Preservative for Sputum Samples. Container: Plastic.
Expand 2 Items
Zenker's fixative solution, Ricca Chemical Company
Supplier: Ricca Chemical
Mercuric Chloride-Potassium Dichromate-Sodium Sulfate Mixture
Expand 2 Items
Cytoseal™ Mounting Medium, Epredia™
Supplier: Epredia
Prevent cracking, fading and discoloration of stained specimens with Epredia™ 60 and 280 mounting media.
Expand 3 Items
Decalcifying Solution for Histology, RICCA Chemical Company
Supplier: Ricca Chemical
Decalcifying Solution, Krajian, for Decalcification of Bones
Expand 2 Items
Xylene Substitute for Processing, Staining and Coverslipping, Epredia
Supplier: Thermo Scientific
Achieve safer, more convenient processing, staining and coverslipping with Epredia™ Xylene Substitute.
Expand 1 Items
Dehydrant Blends, Epredia
Supplier: Thermo Scientific
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
Expand 5 Items
Differentiating solution
Supplier: Thermo Scientific
Epredia™ Differentiation Solution is a standard formulation of hydrochloric acid in denatured alcohol.
Expand 1 Items
Clear-Rite 3®
Supplier: Thermo Scientific
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series™ Clear-Rite 3™, a unique blend of isoparaffinic aliphatic hydrocarbons.