Order Entry
Canada
ContactUsLinkComponent
Tetrahydrofuran-[D₈] (99.5% D) (Isotopic) for NMR spectroscopy
Tetrahydrofuran-[D₈] (99.5% D) (Isotopic) for NMR spectroscopy
Catalog # CAAA36551-03
CAS Number:  
Tetrahydrofuran-[D₈] (99.5% D) (Isotopic) for NMR spectroscopy
Catalog # CAAA36551-03
Supplier Number:  36551-03
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Bottle
  • Size:
    1 g
  • Cat. no.:
    CAAA36551-03
  • Supplier No.:
    36551-03

Specifications

About this item

 :
 :80.04 g/mol
Boiling Pt:66 °C
Melting Pt:–106 °C
Density:0.985
Flash Pt:–17 °C (1 °F)
Storage Temperature:Refrigerator
EINECS:216-898-4
UN:2056
ADR:3,II
 :GHS02
 :GHS07
CAS Number:1693-74-9
Formula:C₄D₈O
MDL Number:MFCD00044238