Order Entry
Canada
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 99+% ACS

L(+)-Ascorbic acid 99+% ACS

L(+)-Ascorbic acid 99+% ACS
Certificates
L(+)-Ascorbic acid 99+% ACS

3 Options Available Below

Product Details & Documents

MDL: MFCD00064328 Beilstein Registry No.: 84272

Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Density: 1.65 Melting Point: ca 191° dec. Beilstein Reference: 84272 Merck Reference: 14,830 MDL No.: MFCD00064328

Synonyms:  Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:14,830
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:∼ 191 °C (dec.)
  • Density:1.65
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Specifications for Product Family

Specification Test Results

  • Specific rotation ¬ +:
    21.0 +/-0.5°
  • Residue after ignition:
    0.1%
  • Heavy Metals (as Pb):
    0.002%
  • Fe:
    0.00001
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $50.72
    $50.72
    Each (25g)
       $50.72
    Minimum order quantity is 1

    Supplier Number: 36237-14
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $84.58
    $84.58
    Each (100g)
       $84.58
    Minimum order quantity is 1

    Supplier Number: 36237-22
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $327.81
    $327.81
    Each (500g)
       $327.81
    Minimum order quantity is 1

    Supplier Number: 36237-36
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.