Sodium hydroxide standardised solution 0.1 N in aqueous solution
About this item
MDL: MFCD00003548
CAS No.: 1310-73-2 Molecular Formula: HNaO Formula Weight: 40.00 Physical Form: Liquid MDL No.: MFCD00003548
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Density:1 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,III
3 Options Available Below
Product Details & Documents
MDL: MFCD00003548
CAS No.: 1310-73-2 Molecular Formula: HNaO Formula Weight: 40.00 Physical Form: Liquid MDL No.: MFCD00003548
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Density:1 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,III
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



