Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Sodium hydroxide 5% (w/v) in aqueous solution

Sodium hydroxide 5% (w/v) in aqueous solution
Certificates
Sodium hydroxide 5% (w/v) in aqueous solution

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00003548

CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Appearance: Colorless UN No.: 1824 MDL No.: MFCD00003548

Synonyms:  Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $46.09
    $46.09
    Each (500ml)
       $46.09

    Supplier Number: 35604-AP
  • Bottle
    4 L
    Log in for availability
    Log in for availability
    Each (4l)
    Each (4l) - $123.36
    $123.36
    Each (4l)
       $123.36

    Supplier Number: 35604-K7

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.