Order Entry
Canada
Orders LinkContactUsLinkComponent
Potassium hydroxide standardised solution 1.0 N in aqueous solution

Potassium hydroxide standardised solution 1.0 N in aqueous solution

Potassium hydroxide standardised solution 1.0 N in aqueous solution
Certificates
Potassium hydroxide standardised solution 1.0 N in aqueous solution

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00003553

CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Physical Form: Liquid Appearance: Colorless UN No.: 1814 MDL No.: MFCD00003553

Synonyms:  Potash caustic, Potassium hydrate, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1814
  • ADR:8,III

Documents

Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.