Orders LinkContactUsLinkComponent
SlideFolder™ Microscope Slide Holders, Simport Scientific
SlideFolder™ Microscope Slide Holders, Simport Scientific
SlideFolder™ Microscope Slide Holders, Simport Scientific
SlideFolder™ Microscope Slide Holders, Simport Scientific
SlideFolder™ Microscope Slide Holders, Simport Scientific
These two-part folders hold up to twenty standard microscope slides
  • All units are stackable
  • Not autoclavable
The high-impact polystyrene base holds slides horizontally with numbered spaces for easy identification. The transparent polystyrene door covers the slides and can be swung behind the folder for space-saving purposes.
Temperature range: –80 to 80°C (–112 to 176°F)
Product Family Options
  • Dimensions
    Color
  • 295×192×11 mm (11 11/16×7 9/16×7/16”)
    Blue
    Case of 10
    Case of 10 - $263.42
    $263.42
    Case of 10
       $263.42
     : M750-20B
  • 295×192×11 mm (11 11/16×7 9/16×7/16”)
    Green
    Case of 10
    Case of 10 - $291.15
    $291.15
    Case of 10
       $291.15
     : M750-20G
  • 295×192×11 mm (11 11/16×7 9/16×7/16”)
    White
    Case of 10
    Case of 10 - $262.14
    $262.14
    Case of 10
       $262.14
     : M750-20W
  • 295×192×11 mm (11 11/16×7 9/16×7/16”)
    Yellow
    Case of 10
    Case of 10 - $266.18
    $266.18
    Case of 10
       $266.18
     : M750-20Y
  • 295×192×11 mm (11 11/16×7 9/16×7/16”)
    Grey
    Case of 10
    Case of 10 - $263.90
    $263.90
    Case of 10
       $263.90
     : M750-20GY
  • 295×192×11 mm (11 11/16×7 9/16×7/16”)
    Assorted
    Case of 10
    Case of 10 - $260.86
    $260.86
    Case of 10
       $260.86
     : M750-20AS
  • 295×192×11 mm (11 11/16×7 9/16×7/16”)
    Opaque/white
    Case of 10
    Case of 10 - $230.13
    $230.13
    Case of 10
       $230.13
     : M752-20WOP