Corning® Reusable Plastic Low Form Beakers, PFA, Corning
Catalog #: 89172-068
Supplier: Corning
Supplier #: 1003P-500
Product available on GSA Advantage®
Log in for availability
About this item
Made of PFA, translucent.
- Excellent chemical resistance
- Good temperature resistance
With a raised graduation scale.
Customers Who Bought This Also Bought