Order Entry
Canada
ContactUsLinkComponent
Leupeptin
Leupeptin
Catalog # 97063-924
Supplier:  VWR
CAS Number:  
undefined
Leupeptin
Catalog # 97063-924
Supplier:  VWR
Supplier Number:  M180-5MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 mg
  • Protein/peptide name:
    Leupeptin
  • Grade:
    Proteomics Grade
  • Cat. no.:
    97063-924
  • Supplier No.:
    M180-5MG

Specifications

About this item

Product is animal-free

  • Activity: Working range: 0.5 µg/ml (1 µM)

Extracted proteins are exposed to the threat of degradation by proteases from the moment cells or tissues are lysed. Proteases present in the living cell are released from their regulated environment into the extract where they may freely degrade their targets, resulting in drastically reduced protein yield and sample
quality. Complex protein solutions are typically treated with a mixture of several protease inhibitors in order to preserve the native state of the proteins.

A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin. Inhibits trypsin, plasmin,papain, kallikrein, thrombin and cathepsin A and B.