You searched for: Hydrometers
Hydrometers measure the specific gravity of liquids or soil grains that are too fine for sieve analysis. The free floating device features a cylindrical stem with a ballast bulb bottom. Weighted with metal instead of lead or mercury, users simply read the graduated measurement at the meniscus. Optically clear for exact conclusions, hydrometers may come calibrated for specific tests. Range subdivisions and accuracy levels differ among the relative density analyzing instruments.
VWR® Soil Analysis ASTM Hydrometers, Traceable to NIST
Supplier: VWR International
Used to determine the specific gravity of soil in the range of 0.995 to 1.038, or -5 to 60 g/L (using a weight by volume scale), these hydrometers conform to the accuracy requirements of ASTM hydrometers.
Expand 2 Items
VWR® Safety Brix Sugar Scale Combined Form Thermo-Hydrometers
Supplier: VWR International
This wide range of combined-form hydrometers are used in the food and beverage, and other industries to measure % sucrose by weight in various ranges from 0° to 61° Brix, and include a convenient, built-in thermometer with a temperature scale of 0/50°C.
Expand 5 Items
VWR® API ASTM Hydrometers, Traceable to NIST
Supplier: VWR International
These plain form hydrometers are available in a variety of models measuring the relative densities (specific gravity) of oil and petroleum products in various ranges from –1° to 96° API in accordance with the American Petroleum Institute (API) and the American Society of Testing Materials (ASTM) E100 standards.
Expand 10 Items
VWR® Alcohol Proof – Ethyl Alcohol Hydrometers, Traceable to NIST
Supplier: VWR International
For use in determining proof ethyl alcohol spirits.
Expand 8 Items
VWR® Specific Gravity ASTM Hydrometers, Traceable to NIST
Supplier: VWR International
Specific gravity range: 1.000 to 1.050. Length: 26cm. With 0.001 divisions.
Expand 14 Items
VWR® Specific Gravity/Relative Density Plain Form Hydrometers, Traceable to NIST
Supplier: VWR International
Specific gravity range: 2.000 to 3.000. Length: 30.5cm. With 0.01 divisions. For liquids considerably heavier than water.
Expand 21 Items
VWR® Universal Specific Gravity and Baume Plain Form Hydrometer, Traceable to NIST
Supplier: VWR International
Universal Hydromete
Expand 1 Items
VWR® Isopropyl Alcohol Hydrometers, Traceable to NIST
Supplier: VWR International
Range 0 to 50%. Length: 26cm. 1% divisions. Measures percent by volume of isopropyl alcohol. Calibrated at 20[degree]C.
Expand 2 Items
VWR® Specific Gravity/Relative Density (g/cm³) Precision Hydrometers, Traceable to NIST
Supplier: VWR International
Available in a wide range of models measuring specific gravity/relative density (g/cmᶟ) of a variety of liquids in various ranges from 0.600 to 1.920, with 0.0005 divisions.
Expand 21 Items
VWR® Brix Sugar Scale Plain Form Hydrometers, Traceable to NIST
Supplier: VWR International
Used in the food and beverage and other industries for measuring % sucrose by weight, these plain form hydrometers are available in various ranges from 0° to 71° Brix.
Expand 8 Items
VWR® Baume Plain Form Hydrometers, Traceable to NIST
Supplier: VWR International
Available in a variety of models measuring Baume of syrups and other liquids heavier than water in various ranges from 0° to 70°, these plain form hydrometers are used in the chemistry, pharmacology, food and beverage, starch industries and more.
Expand 11 Items
VWR® Battery Hydrometer with Siphon Set, Traceable to NIST
Supplier: VWR International
Range: 1.150/1.300 Specific Gravity. 165mm length. 0.002 divisions.
Expand 1 Items
VWR® Specific Gravity/Relative Density and Baume Plain Form Hydrometers, Traceable to NIST
Supplier: VWR International
Used in the chemistry, pharmacology, food and beverage, and starch industries to name a few, these dual scale, plain form hydrometers measure specific gravity / relative density (g/cm3) of liquids lighter and heavier than water in various ranges from 0.700 to 2.00, and Baume in various ranges from 10° to 70°
Expand 10 Items
VWR® Combined Alcohol Proof and Tralles Hydrometer
Supplier: VWR International
Dual scale hydrometer features thermometer in the bottom portion to permit temperature correction of test liquid.
Expand 1 Items
VWR® Salt Brine Plain Form Hydrometers, Traceable to NIST
Supplier: VWR International
Used for measuring sodium chloride saturation of salt brines in various industries including food, fishing, and transportation.
Expand 2 Items
VWR® Calcium Chloride Salometers, Traceable to NIST
Supplier: VWR International
Choose from two models, one measuring percent saturation of CaCl₂ in the range of 0 to 120%, and the other model measuring specific gravity/relative density (g/cmᶟ) from 1.000 to 1.280.
Expand 2 Items
VWR® API ASTM Hydrometers with Individual Calibration Report
Supplier: VWR International
Choose from a wide range of individually calibrated plain form models measuring the relative density (specific gravity) of oil and petroleum related products in various ranges from -1° to 81° API in accordance with the American Petroleum Institute (API) and the American Society of Testing Materials (ASTM) E100 standards.
Expand 7 Items
VWR® ASTM Precision Specific Gravity Hydrometers with Individual Calibration Report, Traceable to NIST
Supplier: VWR International
Available in a variety of individually calibrated models measuring specific gravity of petroleum products and heavy liquids in various ranges from 0.650 to 1.500, in accordance with the American Society of Testing Materials (ASTM) E100 standards
Expand 15 Items
VWR® Specific Gravity Hydrometer Set
Supplier: VWR International
This Hydrometer Set consists of eight DURAC® Specific Gravity/Relative Density (g/cmᶟ) Plain Form Hydrometers for measuring specific gravity of heavy and light liquids in various ranges from 0.700 to 2.000
Expand 1 Items
VWR® DURAC® Safety Specific Gravity/Relative Density (g/cm³) Combined Form Thermo-Hydrometers, Traceable to NIST
Supplier: VWR International
Made of Lead free ballast and glass, these combined form thermo-hydrometers offer the additional convenience of an enclosed thermometer with a temperature scale of 0/150°F, and measure specific gravity / relative density (g/cm3) of a variety of liquids in various ranges from 0.800 to 1.420
Expand 4 Items
Salinity Hydrometer Kit
Supplier: Avantor
Measures Specific Gravity from 1.0–1.07 SG
Expand 1 Items
SP Bel-Art H-B® DURAC® Triple Scale Beer and Wine Hydrometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Achieve perfect beer brewing and wine making results with DURAC Triple Scale Beer and Wine Hydrometers. Includes three scales, Specific Gravity, Potential Alcohol by Volume (PAV), and Brix, that provide invaluable data points when crafting beer, wine, kombucha, mead, cider, and more in either a commercial or home brew setting.
Expand 3 Items
VWR® Safety API Combined Form Thermo-Hydrometer
Supplier: Bel-Art Products, a Part of SP
This combined-form hydrometer, with a convenient, built-in thermometer, measures the relative density (specific gravity) of oil and petroleum related products in accordance with American Petroleum Institute (API) standards.
Expand 1 Items
SP Bel-Art H-B® DURAC® Specific Gravity/Relative Density (g/cm³) Plastic Plain Form Hydrometers, Traceable to NIST, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Expand 5 Items
SP Bel-Art H-B® DURAC® Baume Plastic Plain Form Hydrometers, Traceable to NIST, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Expand 2 Items
SP Bel-Art H-B® DURAC® Salt Brine Plastic Plain Form Hydrometers, Traceable to NIST, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Expand 2 Items
SP Bel-Art H-B® DURAC® Alcohol Proof, Ethyl Alcohol Hydrometers, Traceable to NIST, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Used to determine proof of ethyl alcohol spirits, these hydrometers are available in various ranges.
Expand 2 Items
Brix Hydrometers with Thermometer (°C)
Supplier: Heathrow Scientific
Our brix scale hydrometers with °C thermometer are calibrated to show the percentage of sucrose by weight at 20 °C.
Expand 12 Items
Precision Short Form Specific Gravity Hydrometers
Supplier: Vee Gee Scientific
These precision short form specific gravity hydrometers use less volume of the test liquid while still offering 0.001 resolution.
Expand 7 Items
GOLD BRAND API High Precision Combined Form Hydrometer, Thermco
Supplier: Thermco
High precison API combined form hydrometers made to exceed API specifications, which incorporate a thermometer in the hydrometer