Ribonuclease Inhibitor (RNase Inhibitor)
Catalog # 97062-922
Supplier: VWR
CAS Number:
Specifications
- Size:2 KU
- Protein/peptide name:Ribonuclease Inhibitor (RNase Inhibitor)
- Cat. no.:97062-922
- Supplier No.:E633-2KU
Specifications
About this item
RNase Inhibitor is used to inhibit the activity of RNases in reaction mixtures for cDNA synthesis and in vitro transcription, as well as for long term storage of valuable RNA samples
- Activity: 2kU
A unit of this protein will inhibit 50% of the activity of 5 ng of RNase A.