


Phenol ≥99% saturated solution in water, Biotechnology Grade, buffered
Catalog # 97064-704
Supplier: VWR
CAS Number:
Specifications
- Size:400 ml
- Cat. no.:97064-704
- Supplier No.:0945-400ML
Specifications
Specification Test Results
- Abs.@ 330nm:= 0.2
- Abs.@ 405nm:= 0.1
- Abs.@ 510nm:= 0.1
- pH (Buffer) @25°C:7.7 - 8.1
- Purity:= 99 %
Specification Test Results
About this item
:
:94.11 g/mol
Storage Temperature:Ambient
EINECS:203-632-7
UN:2821
ADR:6.1,II
:GHS07
:GHS05
:GHS08
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00002143