You searched for: Drug Tests
Drug tests offered in various forms accurately analyze biological specimen to detect substance use. Whether examinations include urine, blood, breath, or saliva, these portable tests permit immediate on-site testing. Identifying multiple commonly used parent drugs or metabolities simultaneously, the devices provide rapid and accurate results comparable to laboratories without the excessive costs. Choose among qualitative, semi-qualitative, or quantitative one-step processing drug tests. Multi-test kits and anti-tampering equipment to ensure pure samples are also available.
Clarity Drugs of Abuse Urine Test Cups, Clarity Diagnostics
Supplier: Clarity Diagnostics
Integrated drug panel and testing cup in one.
Expand 3 Items
Clarity FORENSIC Drugs of Abuse Urine Single Dip Test, Clarity Diagnostics
Supplier: Clarity Diagnostics
The Clarity FORENSIC single dip test is a rapid test to detect Drugs of Abuse in urine sample in less than 5 minutes.
Expand 1 Items
Clarity Drugs of Abuse Urine Test Panels, Clarity Diagnostics
Supplier: Clarity Diagnostics
Easy-to-use test panels.
Expand 2 Items
Rapid Response™ Multi-Drug Integrated Split Specimen Cups, BTNX
Supplier: BTNX INC.
The Rapid Response® Multi-drug integrated split specimen test cup is a urine collection device with a built-in qualitative test panel for the simultaneous detection of multiple drugs and metabolites in human urine without specimen handling.
Expand 2 Items
Rapid Response™ Multi-Drug Saliva Test Cups, BTNX
Supplier: BTNX INC.
The Rapid Response® Multi-drug saliva test cup is a rapid, qualitative screening test for the simultaneous detection of multiple drugs and metabolites in human oral fluid. An indicator strip is included in the indicator window of the collection swab to signal saturation. Upon saturation, the collection swab will change colour.
Expand 5 Items
Rapid Response™ Single Drug Test Strip (Urine), BTNX
Supplier: BTNX INC.
The Rapid Response® Single Drug Test Strip Test is a rapid, qualitative test for the detection of a selected drug in human urine.
Expand 4 Items
Rapid Response™ Multi-Drug Test Panels, BTNX
Supplier: BTNX INC.
The Rapid Response® Multi-drug test panel is a rapid, one-step, qualitative test for the simultaneous detection of multiple drugs and metabolites in human urine.
Expand 8 Items
Bluing Tablets, FoxWest Sales
Supplier: Decon Labs
Bluing Tablets are used to color the water in a toilet, via a blue dye, in order to prevent the water from being used to dilute a urine sample for drug testing
Expand 1 Items
Urine Drug Screening, Hydrocodone Assay, MedTest DX
Supplier: MEDTEST DX, INC.
The Hydrocodone assay is intended for the qualitative and semi-quantitative detection and estimation of hydrocodone and its metabolites in human urine at a cutoff of 300 ng/ml.
Expand 2 Items
Rapid Response™ Multi-Drug One Step Cups, BTNX
Supplier: BTNX INC.
The Rapid Response® Multi-drug urine test cup is a rapid, qualitative test for the simultaneous detection of multiple drugs and metabolites in human urine at a specified cut off level.
Expand 5 Items
Rapid Response™ Fentanyl Test Strip Kit (Liquid / Powder), BTNX
Supplier: BTNX INC.
The Rapid Response™ Fentanyl (FYL) forensic test kit is a lateral flow chromatographic immunoassay for the qualitative detection of fentanyl in liquid and powder substances at the cut-off concentration of 200 ng/ml.
Expand 1 Items
Rapid Response™ Multi-Drug Flat Test Cup, BTNX
Supplier: BTNX INC.
The Rapid Response™ multi-drug flat cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs and metabolites in urine samples at specified cut-off levels.
Expand 4 Items
AimTab™ Bluing Tablets, Germaine® Laboratories
Supplier: Germaine Laboratories
Tablets are designed for use in all drug testing collections to prevent adulteration
Expand 1 Items
Rapid Response™ Fentanyl Test Strip, BTNX
Supplier: BTNX INC.
The Rapid Response™ Fentanyl (FYL) Test Strip is a rapid visual immunoassay for the qualitative detection of fentanyl at a cut-off of 200 ng/mL for liquid/powder or 20 ng/mL for urine.
Expand 1 Items
Rapid Response™ Alcohol Test Strip, BTNX
Supplier: BTNX INC.
The Rapid Response® Alcohol test strip is a rapid and highly sensitive method to detect the presence of alcohol in saliva to provide an approximation of relative blood alcohol concentration (BAC) at 0.02% or greater.
Expand 1 Items
Rapid Response™ Lateral Flow Reader, BTNX
Supplier: BTNX INC.
The Rapid Response™ lateral flow reader analyzes results and automatically generates and stores test reports from Rapid Response™ Multi-drug test panels.
Expand 1 Items
AimStrip® Fentanyl Test Strips, Germaine Laboratories
Supplier: Germaine Laboratories
AimStrip® Fentanyl Test Strips are used for the detection of Fentanyl in liquids and powders.
Expand 1 Items
Rapid Response™ Fentanyl Test Strip (Liquid / Powder), BTNX
Supplier: BTNX INC.
The Rapid Response™ Fentanyl Test Strip (Liquid / Powder) detects fentanyl in suspicious substances on surfaces and liquids from suspicious receptables at a cut-off concentration of 200 ng/mL.
Expand 1 Items
Rapid Response™ Xylazine Test Strip (Liquid/Powder), BTNX
Supplier: BTNX INC.
The Rapid Response™ Xylazine Test Strip Kit (Liquid / Powder) detects the presence xylazine in liquid or powder samples at a cut-off concentration of 1000 ng/mL or higher.
Expand 1 Items
AimStrip® Xylazine Urine/Powder Dipstick, Germaine Laboratories
Supplier: Germaine Laboratories
AimStrip® Xylazine Urine/Powder Dipstick, 25/Kit. Cut-off concentration of 1000 ng/ml.