Aprotinin
Supplier: VWR
Certificates
About this item
A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin
GHS Symbols: GHS08
Signal Word: Danger
Not an inhibitor of Factor Xa and thrombin.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.